missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl potassium malonate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC248085000

 View more versions of this product

Catalog No. AC248085000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl potassium malonate
White
500g
6148-64-7
C5H7KO4
MFCD00035603
15,156
Solubility in water: 2.53g/L (25°C)
CCOC(=O)CC(=O)[O-].[K+]
170.2
170.2
98%
Authentic
200.0°C
98%
98%
KOOCCH2COOC2H5
02,III,1609
ethyl malonate potassium salt, ethyl potassium malonate, ethylpotassiummalonate, malonic acid monoethyl ester potassium salt, monoethyl malonate potassium salt, monoethyl potassium malonate, potassium 3-ethoxy-3-oxopropanoate, potassium ethyl malonate, potassium monoethyl malonate, propanedioic acid, monoethyl ester, potassium salt
WVUCPRGADMCTBN-UHFFFAOYSA-M
potassium;3-ethoxy-3-oxopropanoate
3446434
Powder or Cyrstals

Chemical Identifiers

6148-64-7
170.2
WVUCPRGADMCTBN-UHFFFAOYSA-M
3446434
CCOC(=O)CC(=O)[O-].[K+]
C5H7KO4
MFCD00035603
ethyl malonate potassium salt, ethyl potassium malonate, ethylpotassiummalonate, malonic acid monoethyl ester potassium salt, monoethyl malonate potassium salt, monoethyl potassium malonate, potassium 3-ethoxy-3-oxopropanoate, potassium ethyl malonate, potassium monoethyl malonate, propanedioic acid, monoethyl ester, potassium salt
potassium;3-ethoxy-3-oxopropanoate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-156-7

SDS Documents