missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl picolinoylacetate, 95%, Thermo Scientific™

Chemical Identifiers

CAS 26510-52-1
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00094022
InChI Key FQHXWZMJALFSJJ-UHFFFAOYSA-N
Synonym 2-3-ethoxy-3-oxopropanoyl pyridine, d05hbz, ethyl 3-oxo-3-2-pyridinyl propanoate, ethyl 3-oxo-3-2-pyridyl propanoate, ethyl 3-oxo-3-2-pyridyl propionate, ethyl 3-oxo-3-pyridin-2-yl propanoate, ethyl 3-pyridinyl-3-oxopropionate, ethyl picolinoylacetate, ethyl-2-pyridoyl acetate, pubchem11088
PubChem CID 2736461
IUPAC Name ethyl 3-oxo-3-pyridin-2-ylpropanoate
SMILES CCOC(=O)CC(=O)C1=CC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC425470025
SDS View Documents Promotion Details
Thermo Fisher Scientific
425470025
2.5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC425470100
SDS View Documents Promotion Details
Thermo Fisher Scientific
AC425470100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

26510-52-1
193.20
FQHXWZMJALFSJJ-UHFFFAOYSA-N
2736461
CCOC(=O)CC(=O)C1=CC=CC=N1
C10H11NO3
MFCD00094022
2-3-ethoxy-3-oxopropanoyl pyridine, d05hbz, ethyl 3-oxo-3-2-pyridinyl propanoate, ethyl 3-oxo-3-2-pyridyl propanoate, ethyl 3-oxo-3-2-pyridyl propionate, ethyl 3-oxo-3-pyridin-2-yl propanoate, ethyl 3-pyridinyl-3-oxopropionate, ethyl picolinoylacetate, ethyl-2-pyridoyl acetate, pubchem11088
ethyl 3-oxo-3-pyridin-2-ylpropanoate

Specifications

26510-52-1
Brown to Straw-Yellow
>200°C
Authentic
MFCD00094022
Glass bottle
2-3-ethoxy-3-oxopropanoyl pyridine, d05hbz, ethyl 3-oxo-3-2-pyridinyl propanoate, ethyl 3-oxo-3-2-pyridyl propanoate, ethyl 3-oxo-3-2-pyridyl propionate, ethyl 3-oxo-3-pyridin-2-yl propanoate, ethyl 3-pyridinyl-3-oxopropionate, ethyl picolinoylacetate, ethyl-2-pyridoyl acetate, pubchem11088
Solubility in water: negligible
1.164
ethyl 3-oxo-3-pyridin-2-ylpropanoate
2736461
Liquid
Ethyl picolinoylacetate
1.1640g/mL
100°C to 104°C (0.3mmHg)
94% min. (HPLC)
C10H11NO3
22,305
1.5160 to 1.519
2.5g
FQHXWZMJALFSJJ-UHFFFAOYSA-N
CCOC(=O)CC(=O)C1=CC=CC=N1
193.20
193.2
95%
SDS
Product Certifications

RUO – Research Use Only