Learn More
Ethyl p-toluenesulfonate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 147230500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl p-toluenesulfonate | |
32.0°C to 34.0°C | |
1.1740g/mL | |
157°C | |
97.5% min. (GC) | |
C9H12O3S | |
CH3C6H4SO3C2H5 | |
50 g | |
1.174 | |
ethyl p-toluenesulfonate, ethyl tosylate, ethyl p-tosylate, ethyl p-ts, benzenesulfonic acid, 4-methyl-, ethyl ester, ethyl p-methylbenzenesulfonate, ethyl-p-toluenesulfonate, p-toluenesulfonic acid, ethyl ester, ethyl pts, p-toluolsulfonsaeure aethyl ester | |
VRZVPALEJCLXPR-UHFFFAOYSA-N | |
ethyl 4-methylbenzenesulfonate | |
6638 | |
98% |
80-40-0 | |
Colorless to Brown | |
158.0°C to 162.0°C (10.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5100 to 1.5120 | |
MFCD00009100 | |
11, 99 | |
15, 3907 | |
Solubility in water: insoluble. Other solubilities: soluble in many organic solvents | |
CCOS(=O)(=O)C1=CC=C(C=C1)C | |
200.25 | |
200.25 | |
Liquid After Melting |
Chemical Identifiers
80-40-0 | |
200.25 | |
VRZVPALEJCLXPR-UHFFFAOYSA-N | |
6638 | |
CCOS(=O)(=O)C1=CC=C(C=C1)C |
C9H12O3S | |
MFCD00009100 | |
ethyl p-toluenesulfonate, ethyl tosylate, ethyl p-tosylate, ethyl p-ts, benzenesulfonic acid, 4-methyl-, ethyl ester, ethyl p-methylbenzenesulfonate, ethyl-p-toluenesulfonate, p-toluenesulfonic acid, ethyl ester, ethyl pts, p-toluolsulfonsaeure aethyl ester | |
ethyl 4-methylbenzenesulfonate |
Safety and Handling
GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 201-276-7
RUO – Research Use Only