missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl p-toluenesulfonate, 98%, Acros Organics™

Manufacturer:  Acros Organics 147230010

 View more versions of this product

Catalog No. AC147230010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl p-toluenesulfonate
1.1740g/mL
Glass bottle
1.174
158.0°C to 162.0°C (10.0mmHg)
1kg
98%
C9H12O3S
MFCD00009100
15, 3907
VRZVPALEJCLXPR-UHFFFAOYSA-N
ethyl 4-methylbenzenesulfonate
6638
Liquid After Melting
80-40-0
Authentic
1.5100 to 1.5120
32.0°C to 34.0°C
Colorless through Brown
157°C
98%
CH3C6H4SO3C2H5
11, 99
4-Methylbenzenesulfonic acid, ethyl ester, Ethyl p-tosylate
CCOS(=O)(=O)C1=CC=C(C=C1)C
200.252
200.25

Chemical Identifiers

80-40-0
200.252
VRZVPALEJCLXPR-UHFFFAOYSA-N
6638
C9H12O3S
MFCD00009100
4-Methylbenzenesulfonic acid, ethyl ester, Ethyl p-tosylate
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents