Learn More
Ethyl oxalyl chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 118780250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Ethyl oxalyl chloride | |
1.2200g/mL | |
41°C | |
97.5% min. (GC) | |
C4H5ClO3 | |
ClCOCO2C2H5 | |
25 mL | |
1.22 | |
Solubility in water: decomposes. Other solubilities: soluble in common organic solvents | |
CCOC(=O)C(=O)Cl | |
136.53 | |
136.53 | |
Liquid |
4755-77-5 | |
135°C | |
Authentic | |
Glass bottle | |
1.4160 to 1.418 | |
MFCD00000706 | |
02, 541 | |
ethyl chlorooxoacetate, ethyl oxalyl chloride, ethyl chloroglyoxylate, ethoxalyl chloride, ethyl oxalyl monochloride, acetic acid, chlorooxo-, ethyl ester, oxalic acid monoethyl ester chloride, monoethyl oxaloyl chloride, oxalyl chloride, ethyl ester, ethyl chlorocarbonyl formate | |
OWZFULPEVHKEKS-UHFFFAOYSA-N | |
ethyl 2-chloro-2-oxoacetate | |
20884 | |
98% |
Chemical Identifiers
4755-77-5 | |
136.53 | |
OWZFULPEVHKEKS-UHFFFAOYSA-N | |
20884 | |
CCOC(=O)C(=O)Cl |
C4H5ClO3 | |
MFCD00000706 | |
ethyl chlorooxoacetate, ethyl oxalyl chloride, ethyl chloroglyoxylate, ethoxalyl chloride, ethyl oxalyl monochloride, acetic acid, chlorooxo-, ethyl ester, oxalic acid monoethyl ester chloride, monoethyl oxaloyl chloride, oxalyl chloride, ethyl ester, ethyl chlorocarbonyl formate | |
ethyl 2-chloro-2-oxoacetate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 225-285-
RUO – Research Use Only