missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Ethylenedithio)diacetic Acid, TCI America™

Manufacturer:  TCI America E0278100G

 View more versions of this product

Catalog No. E0278100G



Specifications

Specifications

(Ethylenedithio)diacetic Acid
110°C
7244-02-2
MFCD00021771
CCQMORVULNZXIA-UHFFFAOYSA-L
2-[2-(carboxymethylsulfanyl)ethylsulfanyl]acetic acid
310815
Crystal-Powder at 20°C
White
100g
C6H8O4S2
ethylenedithio diacetic acid, 1,2-bis carboxymethylthio ethane, 2-2-carboxymethylsulfanyl ethylsulfanyl acetic acid, 2-2-carboxymethylthio ethylthio acetic acid, 2-carboxymethyl sulfanyl ethyl sulfanyl acetic acid, ethylenedithiodiacetic acid, acmc-2097kv, ethylenebisthio diacetic acid, 1,2-ethylenebis thioglycolic acid, 2,2'-ethylenedithio diacetic acid
C(CSCC(=O)O)SCC(=O)O
208.25
210.26
≥98.0% (GC,T)

Chemical Identifiers

7244-02-2
208.25
CCQMORVULNZXIA-UHFFFAOYSA-L
310815
C(CSCC(=O)O)SCC(=O)O
C6H8O4S2
MFCD00021771
ethylenedithio diacetic acid, 1,2-bis carboxymethylthio ethane, 2-2-carboxymethylsulfanyl ethylsulfanyl acetic acid, 2-2-carboxymethylthio ethylthio acetic acid, 2-carboxymethyl sulfanyl ethyl sulfanyl acetic acid, ethylenedithiodiacetic acid, acmc-2097kv, ethylenebisthio diacetic acid, 1,2-ethylenebis thioglycolic acid, 2,2'-ethylenedithio diacetic acid
2-[2-(carboxymethylsulfanyl)ethylsulfanyl]acetic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents