Please login to your online account to display your discounted pricing

(Ethylenedinitrilo)Tetraacetic Acid, Disodium Salt, Dihydrate, AR, For ACS analysis, Macron Fine Chemicals™


macron

Manufacturer: avantor performance  493120

 View more versions of this product

Catalog No. MK4931A12

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material (Ethylenedinitrilo)Tetraacetic Acid, Disodium Salt, Dihydrate
Name Note ACS
CAS 6381-92-6
Purity Grade 99.0 to 101.0 %
Physical Form Fine white crystalline powder
Molecular Formula C10H14N2Na2O8·2H2O
Formula Weight 372.24g/mol

Specifications

Chemical Name or Material (Ethylenedinitrilo)Tetraacetic Acid, Disodium Salt, Dihydrate
Name Note ACS
CAS 6381-92-6
Purity Grade 99.0 to 101.0 %
Physical Form Fine white crystalline powder
Molecular Formula C10H14N2Na2O8·2H2O
Formula Weight 372.24g/mol