missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenedinitrilo)Tetraacetic Acid ACS AR Powder, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 60-00-4
Physical Form Powder
Molecular Formula C10H16N2O8
Formula Weight 292.25
Chemical Name or Material (Ethylenedinitrilo)Tetraacetic Acid
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK2580500 Avantor Performance
258012
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

60-00-4
C10H16N2O8
(Ethylenedinitrilo)Tetraacetic Acid
Powder
292.25
ACS Grade

Specifications

60-00-4
Powder
292.25
(Ethylenedinitrilo)Tetraacetic Acid
POLYSTORMOR
C10H16N2O8
500g
ACS Grade