Please login to your online account to display your discounted pricing

Ethylenediaminetetraacetic Acid, Trisodiumsalt Hydrate 98%, ACROS Organics™

$103.79 - $103.79

Chemical Identifiers

Chemical Name or Material Ethylenediaminetetraacetic acid, trisodium salt hydrate
Name Note 98%
CAS 85715-60-2
Assay ≥97.5% (corrected for water)
Assay Percent Range 98%
Molecular Formula C10H13N2Na3O8·xH2O
Linear Formula (NaO2CCH2)2NCH2CH2N(CH2CO2Na)CH2CO2H·xH2O
Formula Weight 358.07
MDL Number MFCD00149675
Synonym (Ethylenedinitrilo)tetraacetic acid, trisodium salt hydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC147865000

  SDS
thermo fisher scientific
147865000
Plastic bottle 500g Each for $103.79
Description & Specifications

Specifications

Assay ≥97.5% (corrected for water)
Formula Weight 358.07
Physical Form Crystalline Powder
Quantity 500g
Color White to almost white
Synonym (Ethylenedinitrilo)tetraacetic acid, trisodium salt hydrate
Packaging Plastic bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Water 10% max.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Name Note 98%
Assay Percent Range 98%
CAS 85715-60-2
Linear Formula (NaO2CCH2)2NCH2CH2N(CH2CO2Na)CH2CO2H·xH2O
Molecular Formula C10H13N2Na3O8·xH2O
Chemical Name or Material Ethylenediaminetetraacetic acid, trisodium salt hydrate
MDL Number MFCD00149675

Chelation/Complexation reagent

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.