Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt, 30% w/v aq. soln.

Chelating agent, polymerization catalyst, detergent

$35.11 - $308.89

Chemical Identifiers

Chemical Name or Material Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt
Formula Weight 276.44
MDL Number MFCD01632148
Synonym EDTA tetrakis(tetramethylammonium) salt; Tetrakis(tetramethylammonium) ethylenediaminetetraacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41518AK

  SDS
alfa aesar™
41518AK
250mL Each for $99.14

AA41518AD

  SDS
alfa aesar™
41518AD
50mL Each for $35.11

AA41518K2

  SDS
alfa aesar™
41518K2
1L Each for $308.89
Description & Specifications

Specifications

Formula Weight 276.44
Synonym EDTA tetrakis(tetramethylammonium) salt; Tetrakis(tetramethylammonium) ethylenediaminetetraacetate
Chemical Name or Material Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt
Physical Form Liquid
Concentration 30% w/v aq. soln.
Quantity 250mL
MDL Number MFCD01632148