Learn More
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure, ACROS Organics™
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, C10H12FeN2NaO8.3H2O, CAS Number-18154-32-, sodium ironedetate, edta ferric sodium salt, calmosine, sodium feredetate, edta iron iii sodium salt, sequestrene nafe, ferric sodium edta, ferisan, sodium iron edta, sytron, 1kg
Manufacturer: Thermo Fisher Scientific AC304680051
Description
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, C10H12FeN2NaO8.3H2O, CAS Number-18154-32-, sodium ironedetate, edta ferric sodium salt, calmosine, sodium feredetate, edta iron iii sodium salt, sequestrene nafe, ferric sodium edta, ferisan, sodium iron edta, sytron, 1kg
Chelation/Complexation reagentSpecifications
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate | |
Brown-Yellow | |
98% | |
98% | |
MFCD00798117 | |
calmosine, edta ferric sodium salt, edta iron iii sodium salt, ferisan, ferric sodium edta, sequestrene nafe, sodium feredetate, sodium iron edta, sodium ironedetate, sytron | |
MKWYFZFMAMBPQK-UHFFFAOYSA-J | |
sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+) | |
27461 | |
421.1 | |
98% |
Authentic | |
5kg | |
18154-32-0 | |
C10H12FeN2NaO8·3H2O | |
15,4058 | |
Solubility in water: soluble. | |
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3] | |
421.1 | |
CHEBI:78292 | |
Powder | |
Pure |
Chemical Identifiers
18154-32-0 | |
421.1 | |
MKWYFZFMAMBPQK-UHFFFAOYSA-J | |
27461 | |
sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+) |
C10H12FeN2NaO8·3H2O | |
MFCD00798117 | |
calmosine, edta ferric sodium salt, edta iron iii sodium salt, ferisan, ferric sodium edta, sequestrene nafe, sodium feredetate, sodium iron edta, sodium ironedetate, sytron | |
CHEBI:78292 | |
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3] |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.