Please login to your online account to display your discounted pricing

2,4,6-Triphenyl-s-triazine, 99%

Chemical Identifiers

Chemical Name or Material 2, 4, 6-Triphenyl-s-triazine
Name Note 99%
CAS 493-77-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C21H15N3
Formula Weight 309.36
MDL Number MFCD00006051
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC422190250

  SDS
thermo fisher scientific
422190250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 309.36
Quantity 25g
Color White to off-white
EINECS Number 207-779-8
TSCA TSCA
Melting Point 231° to 236°C
CAS Max % 100.0
Beilstein 26, V,3, 26
CAS Min % 98.5
Synonym TPTZ
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 4, 6-Triphenyl-s-triazine
Name Note 99%
CAS 493-77-6
Molecular Formula C21H15N3
MDL Number MFCD00006051
Assay Percent Range 99%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.