Please login to your online account to display your discounted pricing

Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate (Crystalline Powder/USP/FCC), Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLS312500

 View more versions of this product

Catalog No. S312-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade FCC/USP
Formula Weight 372.24
Melting Point 252°C
Quantity 500g
Physical Form Solid
Assay 99.0 to 101.0% (FCC)
Molecular Formula C10H14N2Na2O8 · 2 H2O
Decomposition Information >252°C
pH 4 to 6
Color White
Identification Pass Test (FCC)
Nitrilotriacetic Acid 0.1% max. (FCC)
Residual Solvents Meets Requirements (USP)
Calcium (Ca) Pass Test (FCC)
MDL Number MFCD00150037
ChemAlert Storage Symbol Gray
Loss on Drying 8.7 to 11.4% (USP)
Packaging Poly Bottle
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:0
Synonym EDTA Disodium Salt
Heavy Metals (as Pb) 0.002% max. (USP)
Lead (Pb) 10mg/kg max. (FCC)
Health Hazard 2 CAUTION!
Chemical Name or Material Ethylenediamine Tetraacetic Acid, Disodium Salt Dihydrate
CAS 6381-92-6

Specifications

Chemical Name or Material Ethylenediamine Tetraacetic Acid, Disodium Salt Dihydrate
CAS 6381-92-6
Purity Grade FCC/USP
Physical Form Solid
Assay 99.0 to 101.0% (FCC)
Molecular Formula C10H14N2Na2O8 · 2 H2O
Formula Weight 372.24
MDL Number MFCD00150037
Synonym EDTA Disodium Salt
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:0

CAUTION!

SDS & Safety Documents