missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate (Crystalline Powder/USP/EP/BP/JP), Fisher Chemical

Manufacturer:  Thermo Scientific S316212

 View more versions of this product

Catalog No. S316212



Specifications

Specifications

Pass Test (BP/EP/JP)
>252°C
37.0 to 39.0% (JP)
Poly Bottle
White
2.5kg
6381-92-6
MFCD00150037
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
44120005
Solid
BP/EP/JP/USP
Ethylenediamine Tetraacetic Acid, Disodium Salt Dihydrate
Pass Test (BP/EP/JP/USP)
8.7 to 11.4 (USP)
Meets Requirements (BP/EP/USP)
252°C
4.0 to 5.5 (BP/EP), 4.3 to 4.7 (JP), 4.0 to 6.0 (USP)
C10H18N2Na2O10
buffer solution, ph 10.00
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]
372.238
372.24
≥99.0% (JP)

Chemical Identifiers

6381-92-6
372.238
OVBJJZOQPCKUOR-UHFFFAOYSA-L
44120005
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]
C10H18N2Na2O10
MFCD00150037
buffer solution, ph 10.00
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:0

SDS Documents