missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for molecular biology, DNAse, RNAse and Protease free

$87.86 - $222.84

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC327201000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
327201000
100 g Plastic bottle
Each for $87.86
 
AC327205000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
327205000
500 g Plastic bottle
Each for $222.84
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent for molecular biology
Specifications

Chemical Identifiers

6381-92-6
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
44120005
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
C10H18N2Na2O10
MFCD00150037,MFCD00003541
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate

Specifications

6381-92-6
252.0°C
4 to 5 (5% soln. at 25°C)
99% min. (Complexometry)
C10H18N2Na2O10
100 g
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
44120005
99+%
0.005% max
Ethylenediaminetetraacetic acid, disodium salt dihydrate
(0.1M in water), (0.1 M in water) at 0nm, 0.02 max. at 280nm, 0.2 max. at 260nm
White
Authentic
Plastic bottle
MFCD00150037,MFCD00003541
153565
Solubility in water: freely soluble. Other solubilities: slightly soluble in alcohol (95%),insoluble in chloroform and ether
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
372.24
372.23
Molecular Biology
Crystalline Solid
Safety and Handling

Safety and Handling

Harmful if inhaled, May cause damage to organs through prolonged or repeated exposure if inhaled

Acute toxicity (category 4), Specific target organ toxicity after repeated exposure (category 2)

EINECSNumber : 200-449-4

RTECSNumber : AH4025000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only