missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediaminetetraacetic Acid Dicalciumsalt 94%, Pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 422130050

 View more versions of this product

Catalog No. AC422130050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Fine Crystalline Powder
White
Authentic
24298-17-7
(CH3(CH2)7)3N(Br)CH2CH2CH3
99%
Trioctylpropylammonium bromide
476.66
5g
74° to 77°C
C27 H58 Br N
Glass bottle
99%
MFCD00041967

Chemical Identifiers

24298-17-7
C27 H58 Br N
476.66
Trioctylpropylammonium bromide, 99%
99%
(CH3(CH2)7)3N(Br)CH2CH2CH3
MFCD00041967
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 246-136-6

TSCA : TSCA