missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediaminetetraacetic Acid Dicalciumsalt 94%, Pure, ACROS Organics™

Chemical Identifiers

CAS 24298-17-7
Assay Percent Range 99%
Molecular Formula C27 H58 Br N
Linear Formula (CH3(CH2)7)3N(Br)CH2CH2CH3
Formula Weight 476.66
MDL Number MFCD00041967
Chemical Name or Material Trioctylpropylammonium bromide, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC422130050 SDS Thermo Fisher Scientific
422130050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Phase transfer catalyst
Description & Specifications

Chemical Identifiers

24298-17-7
C27 H58 Br N
476.66
Trioctylpropylammonium bromide, 99%
99%
(CH3(CH2)7)3N(Br)CH2CH2CH3
MFCD00041967
≥98.5%

Specifications

476.66
5g
74° to 77°C
C27 H58 Br N
Glass bottle
99%
Trioctylpropylammonium bromide, 99%
Fine Crystalline Powder
White
Authentic
24298-17-7
(CH3(CH2)7)3N(Br)CH2CH2CH3
MFCD00041967
≥98.5%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 246-136-6

TSCA : TSCA