missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediaminetetraacetic Acid Calcium Disodium Salt Hydrate, TCI America™

Manufacturer:  TCI America E008825G

 View more versions of this product

Catalog No. E008825G



Specifications

Specifications

Ethylenediaminetetraacetic Acid Calcium Disodium Salt Hydrate
25g
C10H14CaN2Na2O8+2
Ethylenediaminetetraacetic Acid Disodium Calcium Salt, Calcium Disodium Ethylenediaminetetraacetate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.[Na+].[Na+].[Ca+2]
376.286
374.27
≥98.0% (T)
White
62-33-9
MFCD00148919
SHWNNYZBHZIQQV-UHFFFAOYSA-L
calcium;disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate
131664186
Crystal-Powder at 20°C

Chemical Identifiers

62-33-9
376.286
SHWNNYZBHZIQQV-UHFFFAOYSA-L
131664186
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.[Na+].[Na+].[Ca+2]
C10H14CaN2Na2O8+2
MFCD00148919
Ethylenediaminetetraacetic Acid Disodium Calcium Salt, Calcium Disodium Ethylenediaminetetraacetate
calcium;disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1265

RTECSNumber : EV7700000

TSCA : Yes

Documents