missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethylenediaminetetraacetic acid, ACS, 99.4+%

Chelating agent

Manufacturer:  Alfa Aesar™ 1193122

 View more versions of this product

Catalog No. AA1193122



Specifications

Specifications

Ethylenediaminetetraacetic acid
~245°C (decomposition)
60-00-4
(HO2CCH2)2NCH2CH2N(CH2CO2H)2
1716295
edathamil
C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
292.244
CHEBI:42191
Powder
ACS
White
100g
C10H16N2O8
MFCD00003541
14,3517
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
6049
292.24
≥99.4%

Chemical Identifiers

60-00-4
292.244
KCXVZYZYPLLWCC-UHFFFAOYSA-N
6049
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
C10H16N2O8
MFCD00003541
edathamil
CHEBI:42191
C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : AH4025000

TSCA : TSCA

Documents