Learn More
Ethylenediaminetetraacetic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A107130B
Description
Ethylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.
Solubility
Soluble in sodium hydroxide, sodium carbonate, ammonia solution, acid and common organic solvents. Slightly soluble in water.
Notes
Stable. Incompatible with copper, copper alloys, nickel, aluminum, strong oxidizing agents and strong bases.
Specifications
Ethylenediaminetetraacetic acid | |
∼245°C (decomposition) | |
(HO2CCH2)2NCH2CH2N(CH2CO2H)2 | |
1000 g | |
14,3517 | |
Soluble in sodium hydroxide,sodium carbonate,ammonia solution,acid and common organic solvents. Slightly soluble in water. | |
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O | |
292.24 | |
CHEBI:42191 | |
99% |
60-00-4 | |
C10H16N2O8 | |
MFCD00003541 | |
1716295 | |
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol | |
KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid | |
6049 | |
292.24 |
Chemical Identifiers
60-00-4 | |
292.24 | |
KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
6049 | |
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
C10H16N2O8 | |
MFCD00003541 | |
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol | |
CHEBI:42191 | |
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c
H319-H332-H373
EINECSNumber : 200-449-4
RTECSNumber : AH4025000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only