Please login to your online account to display your discounted pricing

Ethylenediamine Tetraacetate, DEPC-Treated Solution (0.5M, pH 8.0), Fisher BioReagents


Quantity 1L
Packaging Nalgene Poly Bottle

Manufacturer: fisher scientific company  BP24831

 View more versions of this product

Catalog No. BP24831

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Quantity 1L
Conductivity [NULL]
Viscosity [NULL]
Color Colorless
pH 8.0
Physical Form Liquid
Concentration 0.475 to 0.525M
Protease Protease free
Merck Index 15, 3565
Chemical Name or Material Ethylenediamine Tetraacetate, DEPC-Treated Solution
Name Note 0.5M, pH 8.0
CAS 60-00-4
ChemAlert Storage Symbol Gray
Health Hazard 2 WARNING!
Identification Pass Test
CAS 1310-73-2
Health Hazard 3 Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:0
Packaging Nalgene Poly Bottle
Recommended Storage RT
CAS 1609-47-8

Specifications

Chemical Name or Material Ethylenediamine Tetraacetate, DEPC-Treated Solution
Name Note 0.5M, pH 8.0
CAS 60-00-4
CAS 1310-73-2
CAS 1609-47-8
Accessories
 

Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

RT

SDS & Safety Documents