missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediamine Tetraacetate, DEPC-Treated Solution (0.5M, pH 8.0), Fisher BioReagents

$371.80 - $395.30

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edathamil, edetic acid, edta, edta acid, endrate, ethylenediaminetetraacetic acid, havidote, sequestrol, titriplex, versene
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP2483100
SDS View Documents Promotion Details
Fisher BioReagents
BP2483100
100mL Poly Bottle
Each for $371.80
Case of 6 Each for $2,230.80
 
BP24831
SDS View Documents Promotion Details
Fisher BioReagents
BP24831
1L Nalgene Poly Bottle
Each for $395.30
Case of 6 Each for $2,371.80
 
Description

Description

EDTA, DEPC-Treated solution is ideal for RNA work.
Specifications

Chemical Identifiers

60-00-4
292.24
KCXVZYZYPLLWCC-UHFFFAOYSA-N
6049
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
C10H16N2O8
MFCD00003541
edathamil, edetic acid, edta, edta acid, endrate, ethylenediaminetetraacetic acid, havidote, sequestrol, titriplex, versene
CHEBI:42191
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

60-00-4,1310-73-2,1609-47-8
MFCD00003541
15, 3565
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
6049
Liquid
Poly Bottle
Colorless
100mL
C10H16N2O8
0.475 to 0.525M
edathamil, edetic acid, edta, edta acid, endrate, ethylenediaminetetraacetic acid, havidote, sequestrol, titriplex, versene
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
292.24
CHEBI:42191
Pass Test
Protease free
8.0
Ethylenediamine Tetraacetate, DEPC-Treated Solution, 0.5M, pH 8.0
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:0

Recommended Storage : RT

SDS
Product Certifications