missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediamine Tetraacetate, DEPC-Treated Solution (0.5M, pH 8.0), Fisher BioReagents

Manufacturer:  Fisher BioReagents BP2483100

 View more versions of this product

Catalog No. BP2483100




Description

Description

EDTA, DEPC-Treated solution is ideal for RNA work.
Specifications

Specifications

Ethylenediamine Tetraacetate, DEPC-Treated Solution
60-00-4,1310-73-2,1609-47-8
0.475 to 0.525M
edathamil, edetic acid, edta, edta acid, endrate, ethylenediaminetetraacetic acid, havidote, sequestrol, titriplex, versene
C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
292.244
CHEBI:42191
Pass Test
Protease free
8.0
0.5M, pH 8.0
C10H16N2O8
15, 3565
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
6049
Liquid
Poly Bottle
Colorless
100mL

Chemical Identifiers

60-00-4
292.244
edathamil, edetic acid, edta, edta acid, endrate, ethylenediaminetetraacetic acid, havidote, sequestrol, titriplex, versene
CHEBI:42191
C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
C10H16N2O8
KCXVZYZYPLLWCC-UHFFFAOYSA-N
6049
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:0

Recommended Storage : RT

SDS Documents