Please login to your online account to display your discounted pricing

Ethylenediamine, Technical Grade, 70% (w/v) ±3% (w/v), LabChem™

$179.17 - $179.17

Chemical Identifiers

Chemical Name or Material Ethylenediamine
CAS 107-15-3
CAS Max % 70
CAS 7732-18-5
CAS Max % 30
Purity Grade Technical Grade
Linear Formula C2H8N2
Formula Weight 60.1
UN Number UN1604
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC141402

 
lab chem
LC141402
Poly Bottle 1L Each for $179.17
Description & Specifications

Specifications

Assay 70% ± 3% (w/v)
Formula Weight 60.1
Odor Ammonia
CAS Max % 30
CAS 107-15-3
Color Clear, colorless to slight yellow
Identification Passes Test
Recommended Storage Room Temperature
CAS Max % 70
CAS 7732-18-5
Physical Form Liquid
Health Hazard 3 GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Health Hazard 2 GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed or in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Purity Grade Technical Grade
Linear Formula C2H8N2
UN Number UN1604
Packaging Poly Bottle
Health Hazard 1 Danger
Chemical Name or Material Ethylenediamine
Quantity 1L
Solubility Information Soluble in water
Safety and Handling

Danger

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed or in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Room Temperature