missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediamine Sulfate, TCI America™

Manufacturer:  TCI America E0082500G

 View more versions of this product

Catalog No. E0082500G



Specifications

Specifications

Ethylenediamine Sulfate
500g
C2H10N2O4S
2811
C(CN)N.OS(=O)(=O)O
158.172
158.17
≥98.0% (T)
White
22029-36-3
MFCD00036417
BNZCDZDLTIHJAC-UHFFFAOYSA-N
ethane-1,2-diamine;sulfuric acid
168015
Crystal-Powder at 20°C

Chemical Identifiers

22029-36-3
158.172
BNZCDZDLTIHJAC-UHFFFAOYSA-N
ethane-1,2-diamine;sulfuric acid
C2H10N2O4S
MFCD00036417
168015
C(CN)N.OS(=O)(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0150

RTECSNumber : KV5425000

TSCA : Yes

Documents