missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediamine Dihydroiodide, TCI America™

Manufacturer:  TCI America E12225G

 View more versions of this product

Catalog No. E12225G



Specifications

Specifications

Ethylenediamine Dihydroiodide
5g
C2H10I2N2
ethylenediamine dihydroiodide, jodethamine, ethane-1,2-diamine dihydroiodide, hydrodine, eddi, ethanediamine dihydroiodide, ethylenediammonium diiodide, unii-721m52nq5l, ethylendiamine dihydroiodide, 1,2-ethanediamine dihydriodide
C(CN)N.I.I
315.925
CHEBI:81719
Crystal-Powder at 20°C
White-Yellow
5700-49-2
MFCD00035535
IWNWLPUNKAYUAW-UHFFFAOYSA-N
ethane-1,2-diamine;dihydroiodide
21921
315.92
≥98.0% (N,T)

Chemical Identifiers

5700-49-2
315.925
IWNWLPUNKAYUAW-UHFFFAOYSA-N
21921
ethane-1,2-diamine;dihydroiodide
C2H10I2N2
MFCD00035535
ethylenediamine dihydroiodide, jodethamine, ethane-1,2-diamine dihydroiodide, hydrodine, eddi, ethanediamine dihydroiodide, ethylenediammonium diiodide, unii-721m52nq5l, ethylendiamine dihydroiodide, 1,2-ethanediamine dihydriodide
CHEBI:81719
C(CN)N.I.I
Safety and Handling

Safety and Handling

TSCA : Yes

Documents