missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediamine Anhydrous (Certified), Fisher Chemical

Manufacturer:  Thermo Scientific E4794

 View more versions of this product

Catalog No. E479-4




Specifications

Specifications

Ethylenediamine Anhydrous
Pass Test
2.07g/mol
2% Max.
Undesignated
4L
107-15-3
C2H8N2
MFCD00008204
ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylendiamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
C(CN)N
60.1
CHEBI:30347
Liquid
Certified
.8980g/cm³
Glass Bottle
12mmHg at 25°C
118°C
8.5°C
11.9
98 to 101 %
NH2CH2CH2NH2
1604
PIICEJLVQHRZGT-UHFFFAOYSA-N
ethane-1,2-diamine
3301
60.1
98 to 101%

Chemical Identifiers

107-15-3
60.1
PIICEJLVQHRZGT-UHFFFAOYSA-N
3301
ethane-1,2-diamine
C2H8N2
MFCD00008204
ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylendiamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
CHEBI:30347
C(CN)N
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents
Accessories