missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediamine, +99%, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC427268000

 View more versions of this product

Catalog No. AC427268000


 

Specifications Description & Specifications

Specifications

Ethylenediamine
0.8980g/mL
AcroSeal™ Glass Bottle
≥99.0%
C2H8N2
H2NCH2CH2NH2
MFCD00008204
0.5% max. (Coulometric)
1, 2-Diaminoethane
800mL
107-15-3
Authentic
Liquid
99+%
1.4555 to 1.4575 (20°C, 589nm)
60.1
1604
15, 3849
Clear

Chemical Identifiers

99+%
H2NCH2CH2NH2
MFCD00008204
Ethylenediamine
C2H8N2
60.1
1, 2-Diaminoethane
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic in contact with skin.
May cause an allergic skin reaction.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger