missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethylenebis(triphenylphosphonium bromide), 98+%

Manufacturer:  Alfa Aesar™ A1219014

 View more versions of this product

Catalog No. AAA1219014



Specifications

Specifications

Ethylenebis (triphenylphosphonium bromide)
≥98%
MFCD00031712
VRKRIHCAYUNAPF-UHFFFAOYSA-L
triphenyl(2-triphenylphosphaniumylethyl)phosphanium;dibromide
2785215
317°C to 320°C
1519-45-5
C38H34Br2P2
3901315
C1=CC=C(C=C1)[P+](CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.[Br-].[Br-]
712.446
712.45
25g

Chemical Identifiers

1519-45-5
712.446
VRKRIHCAYUNAPF-UHFFFAOYSA-L
triphenyl(2-triphenylphosphaniumylethyl)phosphanium;dibromide
C38H34Br2P2
MFCD00031712
2785215
C1=CC=C(C=C1)[P+](CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.[Br-].[Br-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents