Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethylenebis(triphenylphosphonium bromide), 98+%

$116.03 - $391.76

Chemical Identifiers

Chemical Name or Material Ethylenebis (triphenylphosphonium bromide)
CAS 1519-45-5
Molecular Formula C38H34Br2P2
Formula Weight 712.45
MDL Number MFCD00031712
Beilstein 3901315
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1219014

  SDS
alfa aesar™
A1219014
25g Each for $116.03

AAA1219022

  SDS
alfa aesar™
A1219022
100g Each for $391.76
Description & Specifications

Specifications

Formula Weight 712.45
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Ethylenebis (triphenylphosphonium bromide)
Beilstein 3901315
Melting Point 317° to 320°C
MDL Number MFCD00031712
CAS 1519-45-5
Molecular Formula C38H34Br2P2
Assay ≥98%
Health Hazard 1 Warning
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.