Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethylenebis(isothiouronium bromide), 98%

$35.11 - $292.31

Chemical Identifiers

Chemical Name or Material Ethylenebis(isothiouronium bromide)
CAS 6943-65-3
Molecular Formula C4H12Br2N4S2
Formula Weight 340.09
MDL Number MFCD00035055
Beilstein 3914616
Synonym Ethylenediisothiourea dihydrobromide; 2,2'-Ethylenebis(2-thiopseudourea) dihydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0678406

  SDS
alfa aesar™
L0678406
5g Each for $35.11

AAL0678414

  SDS
alfa aesar™
L0678414
25g Each for $96.28

AAL0678422

  SDS
alfa aesar™
L0678422
100g Each for $292.31
Description & Specifications

Specifications

Formula Weight 340.09
Chemical Name or Material Ethylenebis(isothiouronium bromide)
Assay 98%
CAS 6943-65-3
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Molecular Formula C4H12Br2N4S2
MDL Number MFCD00035055
Beilstein 3914616
Synonym Ethylenediisothiourea dihydrobromide; 2,2'-Ethylenebis(2-thiopseudourea) dihydrobromide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Melting Point 224° to 226°C
Quantity 5g
RTECS Number UM6790000
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.