Please login to your online account to display your discounted pricing

Ethylene trithiocarbonate, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC118460250

 View more versions of this product

Catalog No. AC118460250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Low Melting Solid
Formula Weight 136.26
Quantity 25g
Color Yellow
Density 163g/mL
Chemical Name or Material Ethylene trithiocarbonate
Molecular Formula C3H4S3
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
MDL Number MFCD00005493
CAS 822-38-8
Synonym 1, 3-Dithiolane-2-thione

Specifications

Chemical Name or Material Ethylene trithiocarbonate
CAS 822-38-8
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C3H4S3
Formula Weight 136.26
MDL Number MFCD00005493
Synonym 1, 3-Dithiolane-2-thione