Please login to your online account to display your discounted pricing

Ethylene trithiocarbonate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethylene trithiocarbonate
CAS 822-38-8
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C3H4S3
Formula Weight 136.26g/mol
MDL Number MFCD00005493
Synonym 1, 3-Dithiolane-2-thione
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC118460250

  SDS
thermo fisher scientific
118460250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 163g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Quantity 25g
Chemical Name or Material Ethylene trithiocarbonate
CAS 822-38-8
Physical Form Yellow low melting solid
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C3H4S3
Formula Weight 136.26g/mol
MDL Number MFCD00005493
Synonym 1, 3-Dithiolane-2-thione