missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Ethylene Glycol, Technical, Honeywell™

Technical, suitable as anti-freezing agent (with corrosion-inhibitor), blue

$231.76 - $266.54

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044526 View Documents Honeywell™
037701L
1L Poly Bottle Each for $231.76
 
60044527 View Documents Honeywell™
037705L
5L Poly Bottle Each for $266.54
 
Description

Description

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible
Specifications

Chemical Identifiers

107-21-1
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
174
ethane-1,2-diol
C2H6O2
MFCD00002885
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
CHEBI:30742
C(CO)O

Specifications

107-21-1
HOCH2CH2OH
UN3082
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
C(CO)O
62.068
CHEBI:30742
Technical
0.153
Poly Bottle
0.08mmHg (20°C)
111°C (231.8°F)
195°C to 198°C
-20°C
Ethylene Glycol
C2H6O2
MFCD00002885
505945
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
174
62.07g/mol
1.113g/mL at 25°C
752°F
n20/D 1.431(lit.)
2.1 (vs air)
Liquid
Blue
1L
Safety and Handling

Safety and Handling

P260-P301 + P312 + P330

H302-H373

DOTInformation : 9/PGIII