missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, 97%

CAS: 67-42-5 | C14H24N2O10 | 380.35 g/mol

Supplier:  Thermo Scientific Chemicals A1608618

 View more versions of this product

Catalog No. AAA1608618

Only null left
Explore available promotions

Description

Description

Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid is a chelating agent used for making complexes, especially for calcium ions, which help to determine calcium in the presence of magnesium. It can be used experimentally for treating animals possessing cerium poisoning. It is useful in the separation of thorium from monazite. It also acts as a buffer and is used in protein purification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene glycol-O,O′-bis(2-aminoethyl)-N,N,N′,N′-tetraacetic acid is a chelating agent used for making complexes, especially for calcium ions, which help to determine calcium in the presence of magnesium. It can be used experimentally for treating animals possessing cerium poisoning. It is useful in the separation of thorium from monazite. It also acts as a buffer and is used in protein purification.

Solubility
Soluble in 0.1M sodium hydroxide solution. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, alkali, alkali earth metal and many reactive organic and inorganic chemicals.
Specifications

Specifications

Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid
∼242°C (decomposition)
MFCD00004291
1717370
egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
DEFVIWRASFVYLL-UHFFFAOYSA-N
2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
6207
380.35
67-42-5
C14H24N2O10
50 g
14,3529
Soluble in 0.1M sodium hydroxide solution. Slightly soluble in water.
C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
380.35
CHEBI:30740
97%

Chemical Identifiers

67-42-5
380.35
DEFVIWRASFVYLL-UHFFFAOYSA-N
6207
2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
C14H24N2O10
MFCD00004291
egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
CHEBI:30740
C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-651-2

RTECSNumber : AH3760000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only