missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylene Glycol Monoisobutyl Ether 98.0+%, TCI America™
$48.11 - $132.71
Chemical Identifiers
CAS | 4439-24-1 |
---|---|
Molecular Formula | C6H14O2 |
Molecular Weight (g/mol) | 118.176 |
MDL Number | MFCD00059601 |
InChI Key | HHAPGMVKBLELOE-UHFFFAOYSA-N |
Synonym | 2-Isobutoxyethanol, Isobutyl Cellosolve |
PubChem CID | 521158 |
IUPAC Name | 2-(2-methylpropoxy)ethanol |
SMILES | CC(C)COCCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
E030125ML
|
TCI America
E030125ML |
25 mL |
Each for $48.11
|
|
|||||
E0301500ML
|
TCI America
E0301500ML |
500 mL |
Each for $132.71
|
|
|||||
Chemical Identifiers
4439-24-1 | |
118.176 | |
HHAPGMVKBLELOE-UHFFFAOYSA-N | |
521158 | |
CC(C)COCCO |
C6H14O2 | |
MFCD00059601 | |
2-Isobutoxyethanol, Isobutyl Cellosolve | |
2-(2-methylpropoxy)ethanol |
Specifications
4439-24-1 | |
160°C | |
MFCD00059601 | |
1992 | |
HHAPGMVKBLELOE-UHFFFAOYSA-N | |
2-(2-methylpropoxy)ethanol | |
521158 | |
≥98.0% (GC) | |
Ethylene Glycol Monoisobutyl Ether |
Colorless | |
C6H14O2 | |
25 mL | |
2-Isobutoxyethanol, Isobutyl Cellosolve | |
CC(C)COCCO | |
118.176 | |
118.18 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0407&(2)-2424&(7)-0097
RTECSNumber : KL3850000
TSCA : Yes