Please login to your online account to display your discounted pricing

Ethylene Glycol Monoethyl Ether, Reagent Grade, LabChem™

$227.60 - $227.60

Chemical Identifiers

Chemical Name or Material Ethylene Glycol Monoethyl Ether
CAS 110-80-5
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C4H10O2
Formula Weight 90.12
UN Number UN1171
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC141604

 
lab chem
LC141604
Amber Glass 4L Each for $227.60
Description & Specifications

Specifications

Formula Weight 90.12
Chemical Name or Material Ethylene Glycol Monoethyl Ether
Identification Passes Test
EINECS Number 203-804-1
Packaging Amber Glass
Solubility Soluble in water
Storage Information Room Temperature
Purity Grade Reagent Grade
Flash Point 44°C
Viscosity 2.08mPa.s
Decomposition Information Carbon monoxide; Carbon dioxide; Peroxides
Boiling Point 135°C
Physical Form Liquid
Color Clear, colorless
Melting Point −100°C
Quantity 4L
Linear Formula C4H10O2
CAS 110-80-5
UN Number UN1171
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Toxic if inhaled.
May damage fertility or the unborn child.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing vapors.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Contaminated work clothing must not be allowed out of the workplace.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
In case of fire: Use alcohol resistant foam, extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing vapors.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Contaminated work clothing must not be allowed out of the workplace.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
In case of fire: Use alcohol resistant foam, extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Toxic if inhaled.
May damage fertility or the unborn child.