Please login to your online account to display your discounted pricing

Ethylene Glycol Monoethyl Ether (Laboratory), Fisher Chemical

$199.08 - $627.46

Chemical Identifiers

Chemical Name or Material Ethylene Glycol Monoethyl Ether
CAS 110-80-5
Purity Grade Laboratory Grade
Molecular Formula C4H10O2
Linear Formula HOCH2CH2OCH2CH3
Formula Weight 90.12
MDL Number MFCD00002869
Synonym 2-Ethoxyethanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

E180-1

  SDS
fisher scientific company
E1801
Glass Bottle 1L Each for $199.08
Case of 6 Each for $1,194.48

E180-4

  Accessories SDS
fisher scientific company
E1804
Glass Bottle 4L Each for $627.46
Case of 4 Each for $2,509.84
Description & Specifications

Specifications

Formula Weight 90.12
Melting Point −100°C
Boiling Point 135°C
Purity Grade Laboratory Grade
Quantity 1L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Molecular Formula C4H10O2
Color Colorless
pH 7
Vapor Pressure 5mbar at 20°C
Density 0.930g/cm³
Packaging Glass Bottle
Additional Information Vapor Pressure: 5mbar at 20°C
Health Hazard 2 WARNING!
MDL Number MFCD00002869
ChemAlert Storage Symbol Red
Synonym 2-Ethoxyethanol
Identification Pass Test
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Harmful by inhalation, in contact with skin and if swallowed. Irritating to eyes. May cause skin and respiratory tract irritation. May impair fertility. Possible risk
Health Hazard 1 Signal Word:WARNING!
Cleanup Symbol Solvent
Chemical Name or Material Ethylene Glycol Monoethyl Ether
Linear Formula HOCH2CH2OCH2CH3
CAS 110-80-5

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Safety and Handling
 

Signal Word:WARNING!

Emergency Overview
Flammable liquid and vapor. Harmful by inhalation, in contact with skin and if swallowed. Irritating to eyes. May cause skin and respiratory tract irritation. May impair fertility. Possible risk

WARNING!