missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylene Glycol Mono-m-tolyl Ether, TCI America™

Manufacturer:  TCI America E03025ML

 View more versions of this product

Catalog No. E03025ML



Specifications

Specifications

Ethylene Glycol Mono-m-tolyl Ether
Colorless
13605-19-1
MFCD00014035
FDBXUXVQIOQYIX-UHFFFAOYSA-N
2-(3-methylphenoxy)ethanol
83611
Clear Liquid at 20°C
146°C
5mL
C9H12O2
2-3-methylphenoxy ethanol, 2-m-tolyloxy ethanol, ethanol, 2-3-methylphenoxy, ethylene glycol mono-m-tolyl ether, m-cresoxyethanol, 2-3-tolyloxy ethanol, ethanol, 2-m-tolyloxy, 2-3'-methylphenoxy ethanol, ethylene glycol m-cresyl ether, 2-3-methylphenoxy ethan-1-ol
CC1=CC(=CC=C1)OCCO
152.193
152.19
≥98.0% (GC)

Chemical Identifiers

13605-19-1
152.193
FDBXUXVQIOQYIX-UHFFFAOYSA-N
83611
CC1=CC(=CC=C1)OCCO
C9H12O2
MFCD00014035
2-3-methylphenoxy ethanol, 2-m-tolyloxy ethanol, ethanol, 2-3-methylphenoxy, ethylene glycol mono-m-tolyl ether, m-cresoxyethanol, 2-3-tolyloxy ethanol, ethanol, 2-m-tolyloxy, 2-3'-methylphenoxy ethanol, ethylene glycol m-cresyl ether, 2-3-methylphenoxy ethan-1-ol
2-(3-methylphenoxy)ethanol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2777

TSCA : Yes

Documents