missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylene glycol methyl ether acrylate, 98%, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 33516-0010

 View more versions of this product

Catalog No. AC335160010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Ethylene glycol methyl ether acrylate
3121-61-7
Authentic
Liquid
98%
1.4260 to 1.4300 (20°C, 589nm)
40 to 60ppm MEHQ
MFCD00048149
1L
98%
1.0100g/mL
Glass bottle
≥97.5%
C6H10O3
CH3O(CH2)2OC(O)CHCH2
130.14
Clear colorless to slightly yellow

Chemical Identifiers

3121-61-7
C6H10O3
130.14
Ethylene glycol methyl ether acrylate, 98%
98%
CH3O(CH2)2OC(O)CHCH2
MFCD00048149
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Toxic in contact with skin.
May damage fertility. May damage the unborn child.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 221-499-3

RTECSNumber : KL6000000

TSCA : TSCA