Please login to your online account to display your discounted pricing

Ethylene glycol methyl ether acrylate, 98%, stabilized, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC335162500

 View more versions of this product

Catalog No. AC335162500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 130.14
Physical Form Liquid
Assay ≥97.5%
Quantity 250mL
Color Clear colorless to slightly yellow
TSCA TSCA
EINECS Number 221-499-3
RTECS Number KL6000000
Density 1.0100g/mL
Stabilizer 40 to 60ppm MEHQ
Infrared Spectrum Authentic
Refractive Index 1.4260 to 1.4300 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Toxic in contact with skin.
May damage fertility. May damage the unborn child.
Flammable liquid and vapor.
Health Hazard 1 GHS Signal Word: Danger
CAS 3121-61-7
Chemical Name or Material Ethylene glycol methyl ether acrylate
Name Note 98%
Linear Formula CH3O(CH2)2OC(O)CHCH2
Assay Percent Range 98%
MDL Number MFCD00048149
Molecular Formula C6H10O3

Specifications

Chemical Name or Material Ethylene glycol methyl ether acrylate
Name Note 98%
CAS 3121-61-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H10O3
Linear Formula CH3O(CH2)2OC(O)CHCH2
Formula Weight 130.14
MDL Number MFCD00048149
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Toxic in contact with skin.
May damage fertility. May damage the unborn child.
Flammable liquid and vapor.

SDS & Safety Documents