Please login to your online account to display your discounted pricing

Ethylene Glycol (Laboratory), Fisher Chemical


Quantity 20L
Packaging Lined Steel Drum

Manufacturer: fisher scientific company  E17720

 View more versions of this product

Catalog No. E177-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 62.07
Boiling Point 196°C
Purity Grade Laboratory Grade
Melting Point −13°C
Quantity 20L
Physical Form Viscous Liquid
Color Colorless
pH 5.5 to 7.5
Viscosity 21 cP at 20°C
Density 1.113g/cm³
Decomposition Information >500°C
Vapor Pressure 0.12mmHg at 20°C
Packaging Lined Steel Drum
Identification Pass Test
Synonym 1, 2-Ethanediol
Health Hazard 2 WARNING!
MDL Number MFCD00002885
ChemAlert Storage Symbol Gray
Additional Information Vapor Pressure: 0.12mmHg at 20°C
Health Hazard 3 Emergency Overview
Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
Molecular Formula C2H6O2
Linear Formula HOCH2CH2OH
CAS 107-21-1
Chemical Name or Material Ethylene Glycol

Specifications

Chemical Name or Material Ethylene Glycol
CAS 107-21-1
Purity Grade Laboratory Grade
Molecular Formula C2H6O2
Linear Formula HOCH2CH2OH
Formula Weight 62.07
MDL Number MFCD00002885
Synonym 1, 2-Ethanediol
Accessories
 

Emergency Overview
Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0

WARNING!

SDS & Safety Documents