Please login to your online account to display your discounted pricing

Ethylene Glycol (Laboratory), Fisher Chemical

$313.77 - $1,091.38

Chemical Identifiers

Chemical Name or Material Ethylene Glycol
CAS 107-21-1
Purity Grade Laboratory Grade
Molecular Formula C2H6O2
Linear Formula HOCH2CH2OH
Formula Weight 62.07
MDL Number MFCD00002885
Synonym 1, 2-Ethanediol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

E177-4

  Accessories SDS
fisher scientific company
E1774
Glass Bottle 4L Each for $313.77
Case of 4 Each for $1,255.08

E177-20

  SDS
fisher scientific company
E17720
Lined Steel Drum 20L Each for $1,091.38
Description & Specifications

Specifications

Formula Weight 62.07
Melting Point −13°C
Purity Grade Laboratory Grade
Boiling Point 196°C
Quantity 4L
Physical Form Viscous Liquid
Color Colorless
pH 5.5 to 7.5
Density 1.113g/cm³
Viscosity 21 cP at 20°C
Vapor Pressure 0.12mmHg at 20°C
Decomposition Information >500°C
Additional Information Vapor Pressure: 0.12mmHg at 20°C
ChemAlert Storage Symbol Gray
MDL Number MFCD00002885
Packaging Glass Bottle
Synonym 1, 2-Ethanediol
Identification Pass Test
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
Chemical Name or Material Ethylene Glycol
Molecular Formula C2H6O2
CAS 107-21-1
Linear Formula HOCH2CH2OH

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Safety and Handling
 

Emergency Overview
Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0

WARNING!