Please login to your online account to display your discounted pricing

Ethylene Glycol, Fisher BioReagents


Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  BPBP2301

 View more versions of this product

Catalog No. BP230-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Melting Point −13°C
Boiling Point 196°C
Formula Weight 62.06
Quantity 1L
Physical Form Viscous Liquid
pH 5.5 to 7.5
Vapor Pressure 0.12mmHg at 20°C
Density 1.113g/cm³
Color Colorless
Decomposition Information >500°C
Viscosity 21 cP at 20°C
Vapor Density 2.14g/mol
Merck Index 15, 3852
Iron (Fe) 5ppm max.
Identification Pass Test
Assay Percent Range ≥99 %
Additional Information Vapor Pressure: 0.12mmHg at 20°C
Water 0.5% max.
Linear Formula HOCH2CH2OH
Chemical Name or Material Ethylene Glycol
Molecular Formula C2H6O2
Packaging Glass Bottle
Boiling Range 3°C max.
Synonym 1, 2-Ethanediol
Titratable Acid 0.002mEq/g max.
Health Hazard 3 Emergency Overview
Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
Health Hazard 2 WARNING!
ChemAlert Storage Symbol Gray
Recommended Storage RT
CAS 107-21-1

Specifications

Chemical Name or Material Ethylene Glycol
Assay ≥99%
Assay Percent Range ≥99 %
Molecular Formula C2H6O2
Linear Formula HOCH2CH2OH
Formula Weight 62.06
Synonym 1, 2-Ethanediol
Accessories
 

Emergency Overview
Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0

WARNING!

RT

SDS & Safety Documents