Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT, ACROS Organics™
Manufacturer:  Fisher Scientific Acros 118455000
 View more versions of this product
Catalog No. AC118455000
Specifications
Specifications
Ethyleneglycoldimethylether
128-37-0
CH3OCH2CH2OCH3
01,267; 02,143; 16,229
2,5-dioxahexane, dimethoxyethane, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, ethylene dimethyl ether, ethylene glycol dimethyl ether, glycol dimethyl ether, glyme, monoglyme
Liquid
Extra Pure
Authentic
1.3785 to 1.3805
0.867
0.1% max. (K.F.)
1.1 mPa.s (20°C)
Undesignated
6 to 7.5 (1:9)
Stabilized with BHT
99+%
01, 467
15, 3245
90.12
≥99%
0.8670g/mL
Glass bottle
Solubility in water: miscible. Other solubilities: solulbe in hydrocarbon solvents, soluble in ethanol, ethyl ether, acetone, carbon, tetrachloride, benzene, dichloroethane
approx. 100ppm BHT
−6°C
84.0°C to 86.0°C
-69.0°C
500mL
Documents