missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 610350010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethylene glycol dimethyl ether | |
0.8670g/mL | |
Pass Test | |
99% | |
Vapor Pressure: 64hPa at 20°C | |
01, 467 | |
15, 3245 | |
90.12 | |
Extra Dry |
128-37-0 | |
1 L | |
AcroSeal™ Glass Bottle | |
CH3OCH2CH2OCH3 | |
50ppm max. | |
01,267; 02,143; 16,229 | |
monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether | |
≥99% |
RUO – Research Use Only