missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol

Crosslinking monomer

Manufacturer:  Alfa Aesar™ 44151A1

 View more versions of this product

Catalog No. AA44151A1



Specifications

Specifications

Ethylene glycol dimethacrylate
>110°C (230°F)
Light sensitive
Colorless to Yellow
Stabilized with 100ppm 4-methoxyphenol
C10H14O4
MFCD00008590
1,2-bis methacryloyloxy ethane
STVZJERGLQHEKB-UHFFFAOYSA-N
2-(2-methylprop-2-enoyloxy)ethyl 2-methylprop-2-enoate
7355
198.21
98%
1.051 g/mL
1.454
83°C to 85°C (1mmHg)
1kg
97-90-5
H2C=C(CH3)CO2CH2CH2OCOC(CH3)=CH2
1776663
Soluble in water.
CC(=C)C(=O)OCCOC(=O)C(=C)C
198.218
CHEBI:53436
Liquid

Chemical Identifiers

97-90-5
198.218
STVZJERGLQHEKB-UHFFFAOYSA-N
7355
2-(2-methylprop-2-enoyloxy)ethyl 2-methylprop-2-enoate
C10H14O4
MFCD00008590
1,2-bis methacryloyloxy ethane
CHEBI:53436
CC(=C)C(=O)OCCOC(=O)C(=C)C
Safety and Handling

Safety and Handling

GHS H Statement
H317-H335
May cause an allergic skin reaction.
May cause respiratory irritation.

P261-P271-P272-P280g-P302+P352-P304+P340-P312-P333+P313-P363-P501c

H317-H335

EINECSNumber : 202-617-2

RTECSNumber : OZ4400000

TSCA : Yes

Recommended Storage : Keep cold

Documents