missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylene Glycol (Certified), Fisher Chemical

Purity is guaranteed to meet published maximum limits of impurities

Manufacturer:  Thermo Scientific E1781

 View more versions of this product

Catalog No. E178-1




Specifications

Specifications

Ethylene Glycol
0.01% max.
C2H6O2
MFCD00002885
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
C(CO)O
62.068
CHEBI:30742
Viscous Liquid
1.113g/cm³
Glass Bottle
0.12mmHg at 20°C
21 cP at 20°C
196°C
-13°C
1L
107-21-1
Vapor Pressure: 0.12mmHg at 20°C
HOCH2CH2OH
3°C max.
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
174
62.07
Certified
Pass Test
1.112 to 1.115 (at 25°C)
0.2% max.
>500°C
Colorless
5.5 to 7.5

Chemical Identifiers

107-21-1
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
174
ethane-1,2-diol
C2H6O2
MFCD00002885
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
CHEBI:30742
C(CO)O
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0

SDS Documents