Learn More
Ethylene Glycol (Certified), Fisher Chemical™
Purity is guaranteed to meet published maximum limits of impurities
Supplier: Fisher Chemical E178200
Specifications
5.5 to 7.5 | |
-13°C | |
Colorless | |
>500°C | |
196°C | |
Pass Test | |
Viscous Liquid | |
C2H6O2 | |
MFCD00002885 | |
21 cP at 20°C | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
C(CO)O | |
62.068 | |
CHEBI:30742 | |
Certified |
Ethylene Glycol | |
107-21-1 | |
0.01% max. | |
1.113g/cm³ | |
200 L | |
Lined Steel Drum | |
1.112 to 1.115 (at 25°C) | |
HOCH2CH2OH | |
0.12mmHg at 20°C | |
0.2% max. | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
174 | |
62.07 |
Chemical Identifiers
107-21-1 | |
62.068 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
174 | |
ethane-1,2-diol |
C2H6O2 | |
MFCD00002885 | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
CHEBI:30742 | |
C(CO)O |
Safety and Handling
WARNING!
Emergency Overview
Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.
NFPA
Health:2
Flammability:1
Instability:0