missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylene Glycol Bis(3-mercaptopropionate) (Purified), TCI America™

Manufacturer:  TCI America E1329100G

 View more versions of this product

Catalog No. E1329100G



Specifications

Specifications

Ethylene Glycol Bis(3-mercaptopropionate) (Purified)
100g
C8H14O4S2
Ethylene Bis(3-mercaptopropionate), Glycol Bis(3-mercaptopropionate)
C(CS)C(=O)OCCOC(=O)CCS
238.316
238.32
≥98.0% (T)
Yellow
22504-50-3
MFCD00053520
HAQZWTGSNCDKTK-UHFFFAOYSA-N
2-(3-sulfanylpropanoyloxy)ethyl 3-sulfanylpropanoate
89735
Clear Liquid at 20°C

Chemical Identifiers

22504-50-3
238.316
HAQZWTGSNCDKTK-UHFFFAOYSA-N
89735
C(CS)C(=O)OCCOC(=O)CCS
C8H14O4S2
MFCD00053520
Ethylene Bis(3-mercaptopropionate), Glycol Bis(3-mercaptopropionate)
2-(3-sulfanylpropanoyloxy)ethyl 3-sulfanylpropanoate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents